Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZKSCAN2
Entrez Gene ID
702285
Gene Name
zinc finger with KRAB and SCAN domains 2
Gene Aliases
-
Chromosome Location
Chr.20: 23588772 - 23613549 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099539.4 XP_001099539.1 - 3303 100
XM_028841524.1 XP_028697357.1 - 3303 100
GenBank mRNA JV713801.1 - - 398 100

View Full Product Details