Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM9SF1
Entrez Gene ID
715472
Gene Name
transmembrane 9 superfamily member 1
Gene Aliases
EGK_18055
Chromosome Location
Chr.7: 86278277 - 86285203 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266461.1 NP_001253390.1 - 982 143
XM_015143351.2 XP_014998837.1 - 1592 143
XM_015143352.2 XP_014998838.1 - 1607 143
XM_015143353.2 XP_014998839.1 - 1629 143
XM_015143355.2 XP_014998841.1 - 1066 143
XM_028850507.1 XP_028706340.1 - 1225 143
XM_028850508.1 XP_028706341.1 - 1388 143
GenBank mRNA JU320413.1 - - 1098 143
JU470755.1 - - 1271 143
JU470756.1 - - 1256 143
JU474007.1 - - 1256 143
JV045275.1 - - 982 143
JV045276.1 - - 982 143
JV452327.1 - - 977 143
JV635649.1 - - 1127 143

View Full Product Details