Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBWD1
Entrez Gene ID
698333
Gene Name
COBW domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 83672701 - 83726038 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193953.2 NP_001180882.1 - 330 98
XM_028834238.1 XP_028690071.1 - 325 98
XM_028834239.1 XP_028690072.1 - 325 98
XM_028834240.1 XP_028690073.1 - 325 98
GenBank mRNA JU334591.1 - - 279 98
JU470711.1 - - 268 98
JV469294.1 - - 240 98

View Full Product Details