Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB1
Entrez Gene ID
696716
Gene Name
ankyrin repeat and SOCS box containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 125834591 - 125923113 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261538.1 NP_001248467.1 - 116 68
GenBank mRNA JU475966.1 - - 116 68

View Full Product Details