Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
URI1
Entrez Gene ID
700838
Gene Name
URI1 prefoldin like chaperone
Gene Aliases
C19H19orf2
Chromosome Location
Chr.19: 29372383 - 29464740 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085459.4 XP_001085459.3 - 456 104
GenBank mRNA JV664417.1 - - 128 104
JV699930.1 - - 106 104

View Full Product Details