Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
700511
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
EGK_17821
Chromosome Location
Chr.7: 68455590 - 68464896 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001093381.4 XP_001093381.2 - 360 102
GenBank mRNA JU495588.1 - - 138 102

View Full Product Details