Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF17
Entrez Gene ID
706659
Gene Name
fibroblast growth factor 17
Gene Aliases
EGK_18745
Chromosome Location
Chr.8: 22207830 - 22214213 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101524.3 XP_001101524.1 - 535 85

View Full Product Details