Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND5
Entrez Gene ID
711150
Gene Name
BEN domain containing 5
Gene Aliases
-
Chromosome Location
Chr.1: 174445623 - 175900437 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139247.2 XP_014994733.1 - 864 157
XM_015139250.2 XP_014994736.1 - 865 157
XM_015139254.2 XP_014994740.1 - 868 157
XM_015139261.2 XP_014994747.1 - 825 157
XM_015139275.2 XP_014994761.1 - 829 157
GenBank mRNA JV725447.1 - - 758 157

View Full Product Details