Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND5
Entrez Gene ID
711150
Gene Name
BEN domain containing 5
Gene Aliases
-
Chromosome Location
Chr.1: 174445623 - 175900437 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139247.2 XP_014994733.1 - 957 108
XM_015139250.2 XP_014994736.1 - 958 108
XM_015139254.2 XP_014994740.1 - 961 108
XM_015139261.2 XP_014994747.1 - 918 108
XM_015139275.2 XP_014994761.1 - 922 108
GenBank mRNA JV725447.1 - - 851 108

View Full Product Details