Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM241
Entrez Gene ID
700587
Gene Name
transmembrane protein 241
Gene Aliases
EGK_09581
Chromosome Location
Chr.18: 58971729 - 59129351 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015121664.2 XP_014977150.2 - 727 71
XM_015121665.2 XP_014977151.2 - 727 71
XM_028837909.1 XP_028693742.1 - 691 71
XM_028837911.1 XP_028693744.1 - 649 71
XM_028837912.1 XP_028693745.1 - 598 71
XM_028837913.1 XP_028693746.1 - 832 71
XM_028837914.1 XP_028693747.1 - 717 71
GenBank mRNA JV663985.1 - - 705 71

View Full Product Details