Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM241
Entrez Gene ID
700587
Gene Name
transmembrane protein 241
Gene Aliases
EGK_09581
Chromosome Location
Chr.18: 58971729 - 59129351 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015121664.2 XP_014977150.2 - 400 120
XM_015121665.2 XP_014977151.2 - 400 120
XM_028837909.1 XP_028693742.1 - 400 120
XM_028837913.1 XP_028693746.1 - 390 120
GenBank mRNA JV663985.1 - - 378 120

View Full Product Details