Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM69
Entrez Gene ID
712505
Gene Name
tripartite motif containing 69
Gene Aliases
-
Chromosome Location
Chr.7: 21246532 - 21274982 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015142131.2 XP_014997617.2 - 1147 147
XM_015142134.2 XP_014997620.1 - 332 147
XM_028850869.1 XP_028706702.1 - 1147 147

View Full Product Details