Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPLL
Entrez Gene ID
710841
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Gene Aliases
HNRPLL
Chromosome Location
Chr.13: 70316441 - 70359444 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112137.2 XP_014967623.1 - 1180 101
XM_015112138.2 XP_014967624.1 - 1276 101
XM_015112139.2 XP_014967625.1 - 1030 101
XM_015112144.2 XP_014967630.1 - 1188 101
GenBank mRNA JU320592.1 - - 937 101
JU471277.1 - - 1282 101
JU471278.1 - - 1020 101
JV048711.1 - - 937 101

View Full Product Details