Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FEV
Entrez Gene ID
701065
Gene Name
FEV transcription factor, ETS family member
Gene Aliases
-
Chromosome Location
Chr.12: 106414297 - 106418830 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095962.4 XP_001095962.1 - 718 128

View Full Product Details