Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTI2
Entrez Gene ID
696776
Gene Name
TELO2 interacting protein 2
Gene Aliases
C8orf41
Chromosome Location
Chr.8: 33844213 - 33872432 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193802.3 NP_001180731.2 - 1354 156
XM_015145053.2 XP_015000539.1 - 1337 156
GenBank mRNA JU336483.1 - - 994 156
JU472787.1 - - 1093 156
JV044754.1 - - 1117 156

View Full Product Details