Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RILP
Entrez Gene ID
701798
Gene Name
Rab interacting lysosomal protein
Gene Aliases
-
Chromosome Location
Chr.16: 1507411 - 1511260 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086142.4 XP_001086142.1 - 1293 140
XM_028835853.1 XP_028691686.1 - 1287 140
GenBank mRNA JU494491.1 - - 113 140
JV662885.1 - - 336 140

View Full Product Details