Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTPN
Entrez Gene ID
708941
Gene Name
myotrophin
Gene Aliases
EGK_14168
Chromosome Location
Chr.3: 162355682 - 162405132 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260744.1 NP_001247673.1 - 575 84
GenBank mRNA JU336292.1 - - 551 84
JU477125.1 - - 539 84
JU477126.1 - - 539 84
JV048198.1 - - 575 84
JV048199.1 - - 575 84

View Full Product Details