Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKNAD1
Entrez Gene ID
696199
Gene Name
AKNA domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 115403074 - 115444236 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086220.4 XP_001086220.2 - 2506 96
XM_028830849.1 XP_028686682.1 - 2501 96

View Full Product Details