Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H4orf33
Entrez Gene ID
694081
Gene Name
chromosome 5 C4orf33 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 126858453 - 126879972 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JV707081.1 - - 411 128

View Full Product Details