Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICU3
Entrez Gene ID
701957
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
Chromosome Location
Chr.8: 17144079 - 17236996 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028852366.1 XP_028708199.1 - 1297 152
XM_028852367.1 XP_028708200.1 - 1258 152
XM_028852368.1 XP_028708201.1 - 1249 152
XM_028852369.1 XP_028708202.1 - 1201 152
XM_028852370.1 XP_028708203.1 - 1168 152
GenBank mRNA JU326223.1 - - 1030 152
JV614103.1 - - 1204 152

View Full Product Details