Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSMEM1
Entrez Gene ID
702736
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 138755140 - 138765668 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100040.4 XP_001100040.1 - 1034 84

View Full Product Details