Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFT2D2
Entrez Gene ID
699445
Gene Name
SFT2 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 25571311 - 25587796 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265679.1 NP_001252608.1 - 80 124
GenBank mRNA JV635607.1 - - 80 124

View Full Product Details