Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHSY1
Entrez Gene ID
694481
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
-
Chromosome Location
Chr.7: 80463571 - 80538154 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001084554.4 XP_001084554.4 - 330 78
GenBank mRNA JV636035.1 - - 350 78
JV709788.1 - - 67 78

View Full Product Details