Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD17B1
Entrez Gene ID
710392
Gene Name
hydroxysteroid 17-beta dehydrogenase 1
Gene Aliases
-
Chromosome Location
Chr.16: 53685722 - 53690156 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047132.1 NP_001040597.1 - 263 73
XM_028835564.1 XP_028691397.1 - 560 73

View Full Product Details