Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYB5D1
Entrez Gene ID
716429
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
EGK_08149
Chromosome Location
Chr.16: 7650203 - 7706546 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194493.2 NP_001181422.1 - 396 72
GenBank mRNA JU321285.1 - - 415 72
JU472167.1 - - 1054 72
JV044368.1 - - 1029 72
JV663393.1 - - 60 72

View Full Product Details