Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOC8
Entrez Gene ID
713483
Gene Name
exocyst complex component 8
Gene Aliases
EGK_02135
Chromosome Location
Chr.1: 17648985 - 17651581 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258061.1 NP_001244990.1 - 2117 100
GenBank mRNA JU327353.1 - - 1641 100
JU471150.1 - - 2117 100
JU471151.1 - - 2117 100

View Full Product Details