Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC4
Entrez Gene ID
704660
Gene Name
zinc finger DHHC-type palmitoyltransferase 4
Gene Aliases
EGK_13461
Chromosome Location
Chr.3: 39291322 - 39303869 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261639.2 NP_001248568.1 - 513 71
XM_015133238.2 XP_014988724.1 - 772 71
XM_028844900.1 XP_028700733.1 - 772 71
XM_028844901.1 XP_028700734.1 - 886 71
GenBank mRNA JU323399.1 - - 507 71
JV043541.1 - - 513 71

View Full Product Details