The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173469.1 | NP_001166940.1 | 1 - 2 | 858 | 56 |
GenBank mRNA | BC169092.1 | - | 1 - 2 | 858 | 56 |