The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013118.1 | NP_037250.1 | 1 - 2 | 84 | 65 |
GenBank mRNA | M93005.1 | - | 1 - 2 | 84 | 65 |
M95493.1 | - | 1 - 2 | 84 | 65 |