The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019305.2 | NP_062178.1 | 1 - 2 | 705 | 63 |
GenBank mRNA | M22427.1 | - | - | 705 | 63 |
X07285.1 | - | 1 - 2 | 426 | 63 |