The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022260.3 | NP_071596.1 | 5 - 6 | 678 | 65 |
XM_008760546.1 | XP_008758768.1 | 6 - 7 | 1271 | 65 | |
XM_008760547.1 | XP_008758769.1 | 5 - 6 | 1653 | 65 | |
XM_008760548.1 | XP_008758770.1 | 5 - 6 | 980 | 65 | |
XM_008760549.1 | XP_008758771.1 | 6 - 7 | 860 | 65 | |
XM_008760550.1 | XP_008758772.1 | 5 - 6 | 788 | 65 | |
GenBank mRNA | AF072124.1 | - | 5 - 6 | 679 | 65 |
BC070936.1 | - | 5 - 6 | 679 | 65 |