The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053351.1 | NP_445803.1 | 1 - 2 | 246 | 109 |
GenBank mRNA | AF118818.1 | - | 1 - 2 | 246 | 109 |
AF361339.1 | - | 1 - 2 | 213 | 109 |