Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccr5
Entrez Gene ID
117029
Gene Name
chemokine (C-C motif) receptor 5
Gene Aliases
Ckr5, Cmkbr5
Chromosome Location
Chr.8: 133210484 - 133215599 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053960.3 NP_446412.2 1 - 2 58 65
GenBank mRNA Y12009.1 - 1 - 2 68 65

View Full Product Details