The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153468.1 | NP_703198.1 | 4 - 5 | 626 | 71 |
GenBank mRNA | AB082125.1 | - | 4 - 5 | 626 | 71 |