Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psip1
Entrez Gene ID
313323
Gene Name
PC4 and SFRS1 interacting protein 1
Gene Aliases
Ledgf, Psip2, SBP75
Chromosome Location
Chr.5: 101556198 - 101588664 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175765.2 NP_786941.1 1 - 2 175 76
XM_006238353.2 XP_006238415.1 2 - 3 399 76
XM_006238354.2 XP_006238416.1 2 - 3 398 76
XM_008763779.1 XP_008762001.1 2 - 3 398 76
GenBank mRNA AB285525.1 - - 78 76
AF339084.1 - 1 - 2 175 76
AF372090.1 - 1 - 2 175 76
BC093606.1 - 2 - 3 333 76

View Full Product Details