Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mia
Entrez Gene ID
81510
Gene Name
melanoma inhibitory activity
Gene Aliases
Cdrap, Mia1
Chromosome Location
Chr.1: 83991577 - 83993270 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030852.2 NP_110479.1 3 - 4 383 66
GenBank mRNA CA509768.1 - 1 - 2 66
U67884.1 - 3 - 4 383 66

View Full Product Details