The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172065.2 | NP_742062.1 | 3 - 4 | 123 | 96 |
GenBank mRNA | L17318.1 | - | 3 - 4 | 124 | 96 |