Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chp2
Entrez Gene ID
308965
Gene Name
calcineurin-like EF hand protein 2
Gene Aliases
chp-2
Chromosome Location
Chr.1: 192159071 - 192173228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182738.1 NP_877402.1 1 - 2 108 73
XM_008759745.1 XP_008757967.1 2 - 3 286 73
XM_008759746.1 XP_008757968.1 2 - 3 256 73
GenBank mRNA AB086189.1 - 1 - 2 108 73

View Full Product Details