The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032067.1 | NP_114456.1 | 6 - 7 | 1575 | 73 |
GenBank mRNA | BC089788.1 | - | 6 - 7 | 1629 | 73 |
U28830.1 | - | 6 - 7 | 1579 | 73 | |
U82623.1 | - | 6 - 7 | 1575 | 73 |