The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001192002.1 | NP_001178931.1 | 3 - 4 | 346 | 80 |
XM_006245265.2 | XP_006245327.1 | 4 - 5 | 494 | 80 |