The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037159.1 | NP_001032236.1 | - | 2587 | 97 |
GenBank mRNA | AY574027.1 | - | - | 2587 | 97 |