Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh12
Entrez Gene ID
116808
Gene Name
protocadherin 12
Gene Aliases
-
Chromosome Location
Chr.18: 31415533 - 31431108 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053944.1 NP_446396.1 1 - 2 3212 75

View Full Product Details