Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prelid2
Entrez Gene ID
681037
Gene Name
PRELI domain containing 2
Gene Aliases
-
Chromosome Location
Chr.18: 36198522 - 36290796 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271333.1 NP_001258262.1 6 - 7 580 94

View Full Product Details