Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yipf5
Entrez Gene ID
361315
Gene Name
Yip1 domain family, member 5
Gene Aliases
Ac2-256
Chromosome Location
Chr.18: 33401103 - 33436885 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014150.3 NP_001014172.3 1 - 2 89 127
GenBank mRNA BC083754.1 - 1 - 2 89 127

View Full Product Details