Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap19
Entrez Gene ID
679082
Gene Name
Rho GTPase activating protein 19
Gene Aliases
-
Chromosome Location
Chr.1: 261015083 - 261051513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749116.3 XP_003749164.1 1 - 2 76 114

View Full Product Details