Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grsf1
Entrez Gene ID
305256
Gene Name
G-rich RNA sequence binding factor 1
Gene Aliases
-
Chromosome Location
Chr.14: 21039990 - 21056414 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100890.1 NP_001094360.1 8 - 9 1404 122
XM_006250775.2 XP_006250837.1 8 - 9 1291 122
GenBank mRNA BC103654.1 - 3 - 4 375 122

View Full Product Details