Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mthfd2l
Entrez Gene ID
305248
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
Gene Aliases
RGD1310879
Chromosome Location
Chr.14: 18640279 - 18720357 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107211.1 NP_001100681.1 5 - 6 784 121

View Full Product Details