Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mob1b
Entrez Gene ID
360920
Gene Name
MOB kinase activator 1B
Gene Aliases
Mobkl1a
Chromosome Location
Chr.14: 20960052 - 20992753 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108357.1 NP_001101827.1 5 - 6 818 147

View Full Product Details