Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esrrg
Entrez Gene ID
360896
Gene Name
estrogen-related receptor gamma
Gene Aliases
errg
Chromosome Location
Chr.13: 106121602 - 106683353 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203336.2 NP_976081.1 5 - 6 991 59
XM_006250443.2 XP_006250505.1 4 - 5 1644 59
GenBank mRNA AB126962.1 - 4 - 5 875 59
AB126963.1 - 5 - 6 1103 59
AY341057.1 - 4 - 5 754 59

View Full Product Details