Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc4
Entrez Gene ID
362556
Gene Name
tetratricopeptide repeat domain 4
Gene Aliases
-
Chromosome Location
Chr.5: 126264758 - 126282429 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013214.1 NP_001013232.1 3 - 4 402 60
XM_006238519.2 XP_006238581.1 3 - 4 501 60
GenBank mRNA BC083794.1 - 3 - 4 402 60

View Full Product Details